Learn More
Hampton Research HR8-141/CrystalEX Second Generation 96 Well Plate (Corning 3553), 2 ul conical flat drop well, 3 drop well, COC, untreated - 10 plate case
SDP
Catalog No. 500195
Encompass
HR8-141/CrystalEX Second Generation 96 Well Plate (Corning 3553), 2 ul conical flat drop well, 3 drop well, COC, untreated - 10 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-019-5 Supplier Hampton Research Supplier No. HR8141
HR8-141/CrystalEX Second Generation 96 Well Plate (Corning 3553), 2 ul conical flat drop well, 3 drop well, COC, untreated - 10 plate case. Please note items from this supplier are non-returnable.