Learn More
Hampton Research HR8-138/CrystalEX Second Generation 96 Well Plate (Corning 3552), 2 ul round drop well, 3 drop well, COC, untreated - 40 plate case
SDP
Catalog No. 500477
Encompass
HR8-138/CrystalEX Second Generation 96 Well Plate (Corning 3552), 2 ul round drop well, 3 drop well, COC, untreated - 40 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-047-7 Supplier Hampton Research Supplier No. HR8138
Add to Cart
Add to Cart
HR8-138/CrystalEX Second Generation 96 Well Plate (Corning 3552), 2 ul round drop well, 3 drop well, COC, untreated - 40 plate case. Please note items from this supplier are non-returnable.