Learn More
Enzo Life Sciences alphaA/alphaB Crystallin polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA224D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human αA- and αB-Crystallin. UniProt: P02489 (alphaA-crystallin), P02511 (alphaB-crystallin). Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Bovine. Applications: ICC, WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-667-441