Learn More
Enzo Life Sciences alphaB Crystallin polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA223D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of human αB-Crystallin. The sequence is completely conserved in rat, rabbit, and bovine. UniProt: P02511. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Porcine, Bovine. Applications: IF, IHC (PS), WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-667-439