Learn More
Enzo Life Sciences alphaB Crystallin monoclonal antibody (1B6.1 3G4) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA222F
Encompass_Preferred
Clone: 1B6.1-3G4. Host: Mouse. Isotype: IgG1. Immunogen: Bovine αB-crystallin. UniProt ID: P02510. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Chicken, Bovine. Applications: ICC, IF, IHC (PS), WB. Recommended Dilutions: Immunohistochemistry (paraffin sections, 10µg/ml) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7419