Learn More
Sigma Aldrich Fine Chemicals Biosciences crystalline urea 25G
Catalog No. 502907950
Encompass
SRM 912b cert SRM 912b SDSThis Standard Reference Material (SRM) is certified as a neat chemical of known purity. It is a primary standard intended for use during calibration procedures and method validations implemented by clinical laboratories. A unit of SRM 912b consists of 25 g of high-purity crystalline urea.
Catalog No. 50-290-7950 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST912B
SRM 912b cert SRM 912b SDSThis Standard Reference Material (SRM) is certified as a neat chemical of known purity. It is a primary standard intended for use during calibration procedures and method validations implemented by clinical laboratories. A unit of SRM 912b consists of 25 g of high-purity crystalline urea.