Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crystallized Bovine Albumin

Catalog No. AAJ1085606
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ1085606 5 g
AAJ1085609 10 g
AAJ1085622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ1085606 Supplier Thermo Scientific Supplier No. J1085606
Only null left
Add to Cart

Crystallized Bovine Albumin

  • Source: Bovine blood
  • Assay (electrophoresis): ≥99%
  • Identity: By IR
  • Protein: ≥98%
  • pH (7%, 0.15M NaCl): 5.0 - 5.4
  • Moisture (KF) when packaged: %le;5.0%
  • Sodium (Na): 0 - 5.0mg/g
  • Chloride (Cl): 0 - 3.0mg/g
  • CAS: 9048-46-8
TRUSTED_SUSTAINABILITY

Specifications

pH Range 5.0 to 5.4
Quantity 5 g
Source Bovine blood
Packaging Plastic Bottle
Product Type Bovine Albumin
Form Crystalline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.