Learn More
Mitegen 9-Depression Glass Spot Plate, For Crystallization Sitting Drops & Seeding, Cryoprotection Transfers, Heavy Atom Soaks, 1 Plate/Pack
SDP
Catalog No. NC1863768
Encompass
This 9-Depression Glass Spot Plate is an essential, high-quality tool for diverse laboratory applications, especially in crystallography and molecular biology. Featuring nine distinct depressions, this versatile plate facilitates precise, controlled experiments. It is ideally suited for critical tasks including setting up crystallization sitting drops, performing delicate crystallization seeding procedures, and efficient cryoprotection transfers for sensitive samples. Additionally, this durable glass spot plate is perfect for conducting heavy atom soaks, vital for structural determination, and for preparing large drops to produce crystals for advanced neutron diffraction studies. Crafted from high-quality glass, it ensures excellent optical clarity for easy observation and reliable performance through repeated use. Each pack contains one plate, ready to enhance the dependability and efficiency of your laboratory processes.
Catalog No. NC1863768 Supplier Mitegen Supplier No. MC9DGSP
This 9-Depression Glass Spot Plate is an essential, high-quality tool for diverse laboratory applications, especially in crystallography and molecular biology. Featuring nine distinct depressions, this versatile plate facilitates precise, controlled experiments. It is ideally suited for critical tasks including setting up crystallization sitting drops, performing delicate crystallization seeding procedures, and efficient cryoprotection transfers for sensitive samples. Additionally, this durable glass spot plate is perfect for conducting heavy atom soaks, vital for structural determination, and for preparing large drops to produce crystals for advanced neutron diffraction studies. Crafted from high-quality glass, it ensures excellent optical clarity for easy observation and reliable performance through repeated use. Each pack contains one plate, ready to enhance the dependability and efficiency of your laboratory processes.