Learn More
Molecular Dimensions CRYSTALLOPHORE NO1 5X0.6MG
SDP
Catalog No. NC1415670
Encompass_Preferred
Crystallophore No1 (patent pending) is a luminescent Terbium complex that is an effective nucleant and provides significant isomorphous and anomalous phasing power. This is the 5 vial multipack version of this product.
Catalog No. NC1415670 Supplier Molecular Dimensions Supplier No. MD281
Crystallophore No1 (patent pending) is a luminescent Terbium complex that is an effective nucleant and provides significant isomorphous and anomalous phasing power. This is the 5 vial multipack version of this product.