Learn More
Hampton Research HR3-304/CrystalQuick 96 Well Sitting Drop Plate (Greiner 609171), 4 ul square drop well, 1 drop well, low profile - 80 plate case
SDP
Catalog No. 500537
Encompass
HR3-304/CrystalQuick 96 Well Sitting Drop Plate (Greiner 609171), 4 ul square drop well, 1 drop well, low profile - 80 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-053-7 Supplier Hampton Research Supplier No. HR3304
HR3-304/CrystalQuick 96 Well Sitting Drop Plate (Greiner 609171), 4 ul square drop well, 1 drop well, low profile - 80 plate case. Please note items from this supplier are non-returnable.