Learn More
Hampton Research HR3-095G/CrystalQuick Plus 96 Well Sitting Drop Plate (Greiner 609830), 4 ul square drop well, 3 drop well, LBR, hydrophobic - 40 plate case
SDP
Catalog No. 500420
Encompass
HR3-095G/CrystalQuick Plus 96 Well Sitting Drop Plate (Greiner 609830), 4 ul square drop well, 3 drop well, LBR, hydrophobic - 40 plate case. Please note items from this supplier are non-returnable.
Catalog No. 50-042-0 Supplier Hampton Research Supplier No. HR3095G
Add to Cart
Add to Cart
HR3-095G/CrystalQuick Plus 96 Well Sitting Drop Plate (Greiner 609830), 4 ul square drop well, 3 drop well, LBR, hydrophobic - 40 plate case. Please note items from this supplier are non-returnable.