Learn More
Greiner Bio-One CrystalSlide™ Plastic Micro-Channel Platform
Catalog No. 07000550 Shop All Greiner Bio-One Products
Change view
Click to view available options
Plastic Micro-Channel Platform
Catalog No.
07-000-550 Plastic Micro-Channel Platform
1 options
Catalog No. 07-000-550 Supplier Greiner Bio-One Supplier No. 444820
For counter diffusion crystallography. Greiner Bio-One CrystalSlide™ Plastic Micro-Channel Platform features peelable film bottom and has 12 channels per slide.
  • For counter diffusion crystallography
  • Peelable film bottom
  • Peelable film bottom
  • 12 channels per slide
TRUSTED_SUSTAINABILITY
Specifications
Type Plastic Micro-Channel Platform
Description 12 channels/slide
For Use With (Application) Counter Diffusion Crystallography
No. per Case 20