Learn More
Volu Sol Cryptosporidium Stain Kit, 8 oz / 250 mL, Volu-Sol™
SDP
Supplier:  Volu Sol VCK400
Encompass_Preferred
Used to perform the dimethyl sulfoxide acid-fast staining procedure.  Dimethyl sulfoxide (DMSO) is a wetting agent with tremendous penetrating qualities.  When DMSO is mixed with carbol fuchsin, the stain rapidly penetrates cell walls, eliminating the need to heat the slide.  A mild acetic acid is added to the malachite green counterstain combining the decolorization and counterstain steps into one
Catalog No. NC0267000