Learn More
Bioss Crytosporidium parvum
SDP
Catalog No. 5000065991
Encompass_Preferred
Crytosporidium parvum Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65991 Supplier Bioss Supplier No. BSM72087M
Crytosporidium parvum Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide