Learn More
Bioss Crytosporidium parvum
SDP
Supplier:  Bioss BSM72086M
Encompass_Preferred
Crytosporidium parvum Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA IHC Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-66319