Learn More
Aviva Systems Biology Corp CRYZ Antibody - middle region (ARP46065_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46065_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRYZ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CRYZ. Alternate names: DKFZp779E0834, FLJ41475ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59204