Learn More
TARGETMOL CHEMICALS INC CS-722 FREE BASE 5MG
SDP
Catalog No. 502581625
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CS-722 Free base is a synthetic central muscle relaxant with muscle relaxant activity and inhibitory effect on spinal reflex. CS-722 Free base may inhibit spontaneous inhibitory post-synaptic current and excitatory post-synaptic current in hippocampal culture by inhibiting sodium current and calcium current. purity: 97%
Catalog No. 50-258-1625 Supplier TARGETMOL CHEMICALS INC Supplier No. T108935MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CS-722 Free base is a synthetic central muscle relaxant with muscle relaxant activity and inhibitory effect on spinal reflex. CS-722 Free base may inhibit spontaneous inhibitory post-synaptic current and excitatory post-synaptic current in hippocampal culture by inhibiting sodium current and calcium current. purity: 97%