Learn More
BD Cell Analysis 3P CS&T Research Beads
Catalog No. NC1759852
Encompass
D FACSDiva™ CS&T research beads (CS&T research beads) are designed for use on BD™ flow cytometers running BD FACSDiva™ software (v7.0 or later). The beads allow the software to automatically characterize, track, and report measurements of supported BD digital flow cytometers. CS&T research beads are dyed with fluorochromes which are excited by the cytometer’s lasers
Catalog No. NC1759852 Supplier BD Cell Analysis 3P Supplier No. 655051
D FACSDiva™ CS&T research beads (CS&T research beads) are designed for use on BD™ flow cytometers running BD FACSDiva™ software (v7.0 or later). The beads allow the software to automatically characterize, track, and report measurements of supported BD digital flow cytometers. CS&T research beads are dyed with fluorochromes which are excited by the cytometer’s lasers