Learn More
Beckman Coulter Biomek i-Series 230ul Conductive Pipette Tips
Catalog No. NC2178128
Encompass_Preferred
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing
This item is not returnable. View return policy
Catalog No. NC2178128 Supplier Beckman Coulter Supplier No. B85915
Conductive tips for use with Span-8 Pod to enable Liquid Level Sensing