Learn More
Beckman Coulter Biomek i-Series 230 μL Wide-Bore Pipette Tips, Non-Sterile, 960 Pack (10 x 96) - For Biomek Liquid Handlers
Supplier:  Beckman Coulter B85926
Encompass_Preferred
Beckman Coulter Biomek i-Series 230 μL wide-bore pipette tips are disposable tips engineered for automated liquid handling on Biomek platforms. They are non-sterile and supplied in racks to support high-throughput workflows.
  • 230 μL nominal volume
  • Wide-bore geometry for viscous or high-volume dispensing
  • Non-sterile
  • Packaged as 10 racks of 96 tips (960 total)
  • Compatible with 300 μL MC-96 head, 1200 μL MC-96 head, and Span-8 pod
Catalog No. NC2540981