Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B53, with 2 Standard Tubing Ports & 1 Port For Barbed Adapter
Catalog No. 50646532
Encompass
Canary-SafeIDX Waste Cap B53 for Nalgene, 2 Ports for 1/8 in. OD Tubing, 1 Port for Barbed Adabpter (Leak), 1 Port for Charcoal Exhaust Filter
Catalog No. 50-646-532 Supplier Analytical Sales & Services Supplier No. CW53218
Canary-SafeIDX Waste Cap B53 for Nalgene, 2 Ports for 1/8 in. OD Tubing, 1 Port for Barbed Adabpter (Leak), 1 Port for Charcoal Exhaust Filter