Learn More
Analytical Sales & Services Canary-Safe Waste Cap, GL45, 4 Standard Ports, 1 Port For Barbed Adapter
Catalog No. 50646456
Encompass
Canary-SafeIDX Waste Cap GL45, 4 Standard Tubing Ports, 1 Port 6.4mm ID (Leak), 1 Port for Charcoal Exhaust Filter
Catalog No. 50-646-456 Supplier Analytical Sales & Services Supplier No. CW45418
Canary-SafeIDX Waste Cap GL45, 4 Standard Tubing Ports, 1 Port 6.4mm ID (Leak), 1 Port for Charcoal Exhaust Filter