Learn More
Analytical Sales & Services Canary-Safe Waste Cap, GL45, with 2 Standard Tubing Ports & 1 Port For Barbed Adapter
Catalog No. 50646454
Encompass
Canary-SafeIDX Waste Cap GL45, 2 Standard Tubing Ports, 1 Port 6.4mm ID (Leak), 1 Port for Charcoal Exhaust Filter
Catalog No. 50-646-454 Supplier Analytical Sales & Services Supplier No. CW45218
Canary-SafeIDX Waste Cap GL45, 2 Standard Tubing Ports, 1 Port 6.4mm ID (Leak), 1 Port for Charcoal Exhaust Filter