Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ 3500 Dx Genetic Analyzer CS2 Install Kit

Catalog No. 4462042
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 Reactions Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-620-42 1000 Reactions Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-620-42 Supplier Applied Biosystems™ Supplier No. 4462042
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Data Collection software utilizes the multicomponent matrix to automatically correct for the spectral overlap in samples labeled with DS-02 dyes.

The Data Collection software utilizes the multicomponent matrix to automatically correct for the spectral overlap in samples labeled with DS-02 dyes.

Order Info

1 Kit

TRUSTED_SUSTAINABILITY

Specifications

Format Kit
For Use With (Equipment) 3500 Dx Genetic Analyzer
For Use With (Application) Sequencing
Product Type 3500 Dx Genetic Analyzer CS2 Install Kit
Quantity 1000 Reactions Kit
When I started my run on the Applied Biosystems 3500/3500xL Genetic Analyzer, an error occurred: "Unstable Electrophoresis current detected, check for air bubbles" but I don't see any bubbles. What else can cause this?

Some of the other causes of an Unstable Electrophoresis current detected error message are:

1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues

Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.

Do I need to replace the buffer every 14 days if the Applied Biosystems 3500/3500xL Genetic Analyzer is not in use?

If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.