Learn More
Zebra Technologies Corporation Label, Polypropylene, 2x1in (50.8x25.4mm); TT, PolyPro 3000T, Coated, Permanent Adhesive, 1in (25.4mm) core, 2100/roll, 8/box, Plain
Catalog No. NC3706167
Encompass_Preferred
Label, Polypropylene, 2x1in (50.8x25.4mm); TT, PolyPro 3000T, Coated, Permanent Adhesive, 1in (25.4mm) core, 2100/roll, 8/box, Plain
Catalog No. NC3706167 Supplier Zebra Technologies Corporation Supplier No. 17154CS
Label, Polypropylene, 2x1in (50.8x25.4mm); TT, PolyPro 3000T, Coated, Permanent Adhesive, 1in (25.4mm) core, 2100/roll, 8/box, Plain