Learn More
Aviva Systems Biology Corp CSAD Antibody - N-terminal region (ARP64269_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64269_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSAD. It was validated on Western Blot. . Alternate names: CSD, PCAPApplicationWB. Species reactivity: Horse, Human, Mouse, Pig
Catalog No. 50-288-217