Learn More
CSAFE LLC CSafe Global 16L, 120hr, 15°C to 25°C, Controlled-Room-Temperature (CRT) Parcel Shipper - ISTA 7D Qualified - Vacuum Insulation Panel (VIP) Technology. Perfect for long duration shipments of CRT products
Catalog No. NC1866813
Encompass
CSafe Global’s temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance for the safe transport of vital life-science products such as Vaccines, Biologics, Cell Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a 16L payload, qualified to maintain a strict 15°C to 25°C controlled-room-temperature (CRT) condition for 120 hours against ISTA 7D’s worst-case challenge testing. CSafe utilizes proprietary Phase Change Material (PCM) for a simplistic, universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 35L solution is further protected by its double-wall outer corrugate, making this shipper ideal as a reusable system; while also being cost effective for one-way single-use shipments.
Catalog No. NC1866813 Supplier CSAFE LLC Supplier No. A16C120000U
CSafe Global’s temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance for the safe transport of vital life-science products such as Vaccines, Biologics, Cell Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a 16L payload, qualified to maintain a strict 15°C to 25°C controlled-room-temperature (CRT) condition for 120 hours against ISTA 7D’s worst-case challenge testing. CSafe utilizes proprietary Phase Change Material (PCM) for a simplistic, universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 35L solution is further protected by its double-wall outer corrugate, making this shipper ideal as a reusable system; while also being cost effective for one-way single-use shipments.