Learn More
CSAFE LLC CSafe Global 16L, 120hr Frozen Parcel Shipper - ISTA 7D Qualified - Vacuum Insulation Panel (VIP) Technology. Perfect for long duration shipments of Vaccines, Biologics and frozen therapies; including COVID-19 shipments
Catalog No. NC1866818
Encompass
CSafe Global’s temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance and versality for the safe transport of vital life-science products such as Vaccines, Biologics, Cell & Gene Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a payload capacity of 16L and has been qualified to maintain frozen temperatures for 120 hours against ISTA 7D’s worst-case challenge testing, utilizing a universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 16L solution is further protected by its double-wall outer corrugate, making this solution ideal as a reusable system; while also being cost effective for one-way single-use shipments.
Catalog No. NC1866818 Supplier CSAFE LLC Supplier No. A16F120000U
CSafe Global’s temperature-controlled parcel shipper solutions are constructed using best-in-class Vacuum Insulated Panel (VIP) technology, assuring greater peace of mind in your cold chain. These shippers provide industry-leading temperature management performance and versality for the safe transport of vital life-science products such as Vaccines, Biologics, Cell & Gene Therapies, and Oncology Treatments to distribution centers, healthcare facilities and direct to patients.This shipping solution has a payload capacity of 16L and has been qualified to maintain frozen temperatures for 120 hours against ISTA 7D’s worst-case challenge testing, utilizing a universal pack-out. This system has also passed ISTA 3A Distribution Testing.This 16L solution is further protected by its double-wall outer corrugate, making this solution ideal as a reusable system; while also being cost effective for one-way single-use shipments.