Learn More
Molecular Dimensions CSalt Screen HT-96 Block, 96 x 1 mL
SDP
Catalog No. NC3456997
Encompass
CSalt is a 96-condition preformulated sparse matrix screen utilizing a broad array of salt-only solutions of varying concentrations and pH. MD1-141 is presented as 96 x 1 mL conditions.Features:Salt-and-pH-only sparse matrix crystallisation screenWide pH range, 4.6-8.522 unique salts at varying concentrations
Catalog No. NC3456997 Supplier Molecular Dimensions Supplier No. MD1141
CSalt is a 96-condition preformulated sparse matrix screen utilizing a broad array of salt-only solutions of varying concentrations and pH. MD1-141 is presented as 96 x 1 mL conditions.Features:Salt-and-pH-only sparse matrix crystallisation screenWide pH range, 4.6-8.522 unique salts at varying concentrations