Learn More
BD CSampler™ Plus Loader
Catalog No. BDB660519 Shop All BD Biosciences Products
Change view
Click to view available options
For Use With (Equipment):
BD Accuri C6 Plus Flow Cytometer
Catalog No. For Use With (Equipment)
BDB660519 BD Accuri C6 Plus Flow Cytometer
1 options
This item is not returnable. View return policy
Catalog No. BDB660519 Supplier BD Supplier No. 660519
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
BD CSampler Plus, 24-tube rack and 96-well plate automated sample loader for the BD Accuri C6 Plus Flow Cytometer.
  • For the BD Accuri C6 Plus Flow Cytometer
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) BD Accuri C6 Plus Flow Cytometer
WARNING: Reproductive Harm - www.P65Warnings.ca.gov