Learn More
Cell Systems Corporation Attachment Factor - 100mL (4Z0-210)
SDP
Supplier:  Cell Systems Corporation 4Z0210
Encompass_Preferred
Attachment Factor (10mL) is a 5-second coating solution that promotes cell attachment to tissue culture surfaces and encourages correct polarity and cytoskeletal organization. It is vital when cultures are to be initiated, grown, passaged, or used within the Cell Systems Media family. It enhances plating efficiency and growth rates, shortens "lag time" after passage, promotes the organization of polarity and cytoskeletal architecture, and can increase cell yield during culture expansion. Unlike other coating solutions that require long wait time (such as collagen), Attachment Factor provides an ideal coating in just 5 seconds. Use with all except Melanocyte and Keratinocyte cells. Available in 10mL (4Z0-201) and 100mL. Large quantities? Contact Fisher Scientific sales rep to discuss bulk order needs. Tight deadline? Available for next day delivery after the purchase order is received. Maximize results!
Catalog No. 50-900-033