Learn More
Cell Systems Corporation Cell Freezing Medium (4Z0-705)
SDP
Catalog No. 50900041
Encompass_Preferred
Our Cell Freezing Medium (50 mL) is a specialized medium providing the optimal environment for the freeze/thaw cycle of cell cultures and assists in minimizing cellular damage during the stressful process. Use it in conjunction with Passage Reagent Group (PRG) and Attachment Factor for optimal post-thaw protection and cell viability. Use of the Passage Reagent Group (PRG) to release cells from adherent culture for freezing is critically important in minimizing membrane and cytoskeletal damage. Use of PRG in conjunction with Cell Freezing Medium and Attachment Factor minimizes cellular stress and damage associated with these processes and ensures improved viability post-thaw. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent Group 4Z0-800), both available for purchase.
Catalog No. 50-900-041 Supplier Cell Systems Corporation Supplier No. 4Z0705
Our Cell Freezing Medium (50 mL) is a specialized medium providing the optimal environment for the freeze/thaw cycle of cell cultures and assists in minimizing cellular damage during the stressful process. Use it in conjunction with Passage Reagent Group (PRG) and Attachment Factor for optimal post-thaw protection and cell viability. Use of the Passage Reagent Group (PRG) to release cells from adherent culture for freezing is critically important in minimizing membrane and cytoskeletal damage. Use of PRG in conjunction with Cell Freezing Medium and Attachment Factor minimizes cellular stress and damage associated with these processes and ensures improved viability post-thaw. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent Group 4Z0-800), both available for purchase.