Learn More
Cell Systems Corporation Complete Classic Medium With Serum and CultureBoost (4Z0-500)
SDP
Catalog No. 50900036
Encompass_Preferred
Our best-selling medium is optimized for most traditional cell culture experiments. This medium is formulated with 10% serum and becomes a complete medium once activated with the included CultureBoost supplement. This kit consists of 500mL Cell Systems Medium, 5mL CultureBoost (containing animal derived growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). 4Z0-500 is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to use with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
Catalog No. 50-900-036 Supplier Cell Systems Corporation Supplier No. 4Z0500
Our best-selling medium is optimized for most traditional cell culture experiments. This medium is formulated with 10% serum and becomes a complete medium once activated with the included CultureBoost supplement. This kit consists of 500mL Cell Systems Medium, 5mL CultureBoost (containing animal derived growth factors) and 10mL Attachment Factor (extracellular matrix reagent for coating flasks). 4Z0-500 is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to use with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.