Learn More
Cell Systems Corporation Complete Serum-Free Medium Kit With RocketFuel (SF-4Z0-500)
SDP
Catalog No. 50900052
Encompass_Preferred
Our serum-free medium becomes a chemically defined complete medium once activated with the included RocketFuel supplement. This kit consists of 500mL Serum-Free Medium, 8mL RocketFuel (containing animal derived growth factors) and 10mL vial of Attachment Factor (containing animal derived growth factors). SF-4Z0-500 is certified and intended for experimental application. It is bovine-free, chemically defined medium with albumin and phenol red. This media is with no added hormones or antibiotics for optimized experimental conditions. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase. For a bovine-free supplement, see Complete Serum-Free Medium Kit With Recombinant RocketFuel-R (SF-4Z0-500-R).
Catalog No. 50-900-052 Supplier Cell Systems Corporation Supplier No. SF4Z0500
Our serum-free medium becomes a chemically defined complete medium once activated with the included RocketFuel supplement. This kit consists of 500mL Serum-Free Medium, 8mL RocketFuel (containing animal derived growth factors) and 10mL vial of Attachment Factor (containing animal derived growth factors). SF-4Z0-500 is certified and intended for experimental application. It is bovine-free, chemically defined medium with albumin and phenol red. This media is with no added hormones or antibiotics for optimized experimental conditions. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase. For a bovine-free supplement, see Complete Serum-Free Medium Kit With Recombinant RocketFuel-R (SF-4Z0-500-R).