Learn More
Cell Systems Corporation CultureBoost (4CB-500)
SDP
Catalog No. 50900029
Encompass_Preferred
CultureBoost is our most popular growth factor for use in cell biology experiments. Our high-quality formula, containing bovine growth factor and porcine heparin, is suitable for caring for extremely fastidious cells. Use of CultureBoost can result in as much as 25% increase in recovery from freezing, an increase of 10-20% in cell count per passage, and a one day decrease in time-to-confluence. CultureBoost is qualified for use with Cell Systems media. It is available in a 10mL vial in several Cell Systems media kits and "The System" as well as for purchase separately. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800).
Catalog No. 50-900-029 Supplier Cell Systems Corporation Supplier No. 4CB500
CultureBoost is our most popular growth factor for use in cell biology experiments. Our high-quality formula, containing bovine growth factor and porcine heparin, is suitable for caring for extremely fastidious cells. Use of CultureBoost can result in as much as 25% increase in recovery from freezing, an increase of 10-20% in cell count per passage, and a one day decrease in time-to-confluence. CultureBoost is qualified for use with Cell Systems media. It is available in a 10mL vial in several Cell Systems media kits and "The System" as well as for purchase separately. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Available for next day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800).