Learn More
Cell Systems Corporation Medium Kit With Serum and Without Growth Factor (4Z0-500-S)
SDP
Catalog No. 50900038
Encompass_Preferred
Our growth factor-free complete medium is formulated with 10% serum. You can customize it to fit your cell culture needs by completing it CultureBoost growth supplement (4CB-500) or own lab equivalent. This kit consists of 500mL Growth Factor-Free Medium and 10mL Attachment Factor. 4Z0-500-S is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.
Catalog No. 50-900-038 Supplier Cell Systems Corporation Supplier No. 4Z0500S
Our growth factor-free complete medium is formulated with 10% serum. You can customize it to fit your cell culture needs by completing it CultureBoost growth supplement (4CB-500) or own lab equivalent. This kit consists of 500mL Growth Factor-Free Medium and 10mL Attachment Factor. 4Z0-500-S is certified and intended for experimental application. The media is phenol-red free with no added hormones or antibiotics for optimized experimental conditions. It is certified to be used with more than 100 human and animal cell cultures and cell lines. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Research on a tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase.