Learn More
Cell Systems Corporation PRG-3 Trypsin Inhibitor Solution (4Z0-410)
SDP
Catalog No. 50900035
Encompass_Preferred
The PRG-3 Trypsin Inhibitor-dPBS (4Z0-410) is the final step in our Passage Reagent Group (PRG) system, a matched set of reagents optimized for releasing cells from culture dishes and flasks in subculture or freezing procedures. Its primary objective is to ensure cell viability, maintain the integrity of cell surface markers and minimize stress to cells during the detachment process. Available individually or as part of the PRG (4Z0-800) or Passage Reagent System (4Z0-840). The EDTA in PRG-1 loosens cell attachment gently from culture surfaces. Trypsin concentration in PRG-2 is optimized to detach loosened cells while minimizing adverse effects on cell viability and surface markers. Trypsin inhibitor in PRG-3 promptly halts trypsin activity once cells have been successfully released. Bulk order? Contact Fisher Scientific sales rep. Tight deadline? Available for next day delivery after the purchase order is received. Try our primary human cells: ideal for use with media & reagents.
Catalog No. 50-900-035 Supplier Cell Systems Corporation Supplier No. 4Z0410
The PRG-3 Trypsin Inhibitor-dPBS (4Z0-410) is the final step in our Passage Reagent Group (PRG) system, a matched set of reagents optimized for releasing cells from culture dishes and flasks in subculture or freezing procedures. Its primary objective is to ensure cell viability, maintain the integrity of cell surface markers and minimize stress to cells during the detachment process. Available individually or as part of the PRG (4Z0-800) or Passage Reagent System (4Z0-840). The EDTA in PRG-1 loosens cell attachment gently from culture surfaces. Trypsin concentration in PRG-2 is optimized to detach loosened cells while minimizing adverse effects on cell viability and surface markers. Trypsin inhibitor in PRG-3 promptly halts trypsin activity once cells have been successfully released. Bulk order? Contact Fisher Scientific sales rep. Tight deadline? Available for next day delivery after the purchase order is received. Try our primary human cells: ideal for use with media & reagents.