Learn More
Cell Systems Corporation Serum-Free Medium Kit Without Growth Factor (SF-4Z0-500-S)
SDP
Catalog No. 50900054
Encompass_Preferred
Our base medium, which is free of serum and growth factors, can be customized to meet your specific cell culture needs. You can customize it by completing it with serum-free growth factors such as RocketFuel or RocketFuel-R. This kit consists of 500mL Serum-Free Medium, 10mL Attachment Factor (matric reagent for coating flask) and 8mL RocketFuel-S). SF-4Z0-500-S is certified and intended for experimental application. This albumin and phenol red-supplemented media is formulated without added hormones or antibiotics for optimized experimental conditions. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase. For a bovine-free supplement, see Complete Serum-Free Medium Kit With Recombinant RocketFuel-R (SF-4Z0-500-R).
Catalog No. 50-900-054 Supplier Cell Systems Corporation Supplier No. SF4Z0500S
Our base medium, which is free of serum and growth factors, can be customized to meet your specific cell culture needs. You can customize it by completing it with serum-free growth factors such as RocketFuel or RocketFuel-R. This kit consists of 500mL Serum-Free Medium, 10mL Attachment Factor (matric reagent for coating flask) and 8mL RocketFuel-S). SF-4Z0-500-S is certified and intended for experimental application. This albumin and phenol red-supplemented media is formulated without added hormones or antibiotics for optimized experimental conditions. Large quantities? Contact your Fisher Scientific sales representative to discuss your bulk order needs. Tight deadline? Our products are available for next-day delivery after the purchase order is received. Maximize your results! Try our Primary Human Cells and Passage Reagent (4Z0-800), both available for purchase. For a bovine-free supplement, see Complete Serum-Free Medium Kit With Recombinant RocketFuel-R (SF-4Z0-500-R).