Learn More
Aviva Systems Biology Corp CSDA Antibody - C-terminal region (P100917_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100917_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CSDA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CSDA. Alternate names: DBPA, CSDA1, ZONABApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53806