Learn More
Aviva Systems Biology Corp CSDC2 Antibody - N-terminal region (ARP40768_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40768_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CSDC2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CSDC2. Alternate names: PIPPIN, dJ347H13.2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57619