Learn More
Aviva Systems Biology Corp CSEN Antibody - N-terminal region (ARP31932_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31932_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSEN. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CSEN. Alternate names: CSEN, DREAM, KCHIP3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54379