Learn More
Aviva Systems Biology Corp CSF1 Antibody - middle region (ARP44330_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44330_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CSF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CSF1. Alternate names: MCSF, MGC31930, CSF-1ApplicationIF, WB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-58783