Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Csf1r-in-1 | 2095849-04-8 | 98.0% | 479.45 g/mol | C25H20F3N5O2 | 10 MG
SDP

Catalog No. 5000245784
Encompass_Preferred

CSF1R-IN-1 is a potent small-molecule inhibitor of colony stimulating factor 1 receptor (CSF1R) with sub-nanomolar cellular potency. It is supplied for research use to probe CSF1R signaling, macrophage biology, and related disease models.

  • Potent CSF1R inhibition with reported IC50 of 0.5 nM.
  • High purity: 98.04%.
  • Molecular formula C25H20F3N5O2 and molecular weight 479.45 g/mol.
  • Available in powder and ready-to-use DMSO solutions; common pack sizes include 10 mg.
  • Storage: powder at -20 °C; in solvent at -80 °C (up to 6 months) or -20 °C (up to 1 month).
  • For research use only, not intended for human or veterinary use.

Catalog No. 50-002-45784 Supplier Medchemexpress LLC Supplier No. HY10177410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CSF1R-IN-1 is a potent small-molecule inhibitor of colony stimulating factor 1 receptor (CSF1R) with sub-nanomolar cellular potency. It is supplied for research use to probe CSF1R signaling, macrophage biology, and related disease models.

  • Potent CSF1R inhibition with reported IC50 of 0.5 nM.
  • High purity: 98.04%.
  • Molecular formula C25H20F3N5O2 and molecular weight 479.45 g/mol.
  • Available in powder and ready-to-use DMSO solutions; common pack sizes include 10 mg.
  • Storage: powder at -20 °C; in solvent at -80 °C (up to 6 months) or -20 °C (up to 1 month).
  • For research use only, not intended for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.