Learn More
Medchemexpress LLC CSF1R-IN-1 | 2095849-04-8 | 98.0% | 479.45 g/mol | C25H20F3N5O2 | 25 MG
SDP
Catalog No. 5000245704
Encompass_Preferred
CSF1R-IN-1 is a potent small-molecule inhibitor of the colony stimulating factor 1 receptor (CSF1R) used for research into macrophage biology, tumor-associated macrophages, cancer, inflammation, and neurodegeneration. The compound has an IC50 of 0.5 nM against CSF1R, a molecular weight of 479.45 g/mol, and is supplied as a white to off-white solid with reported purity of 98.04%.
  • Potent CSF1R inhibition (IC50 0.5 nM).
  • High purity suitable for research (≈98.0%).
  • Defined molecular properties: C25H20F3N5O2, 479.45 g/mol.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage: powder -20°C (up to 3 years).
  • Available in small research quantities including 25 MG.
Catalog No. 50-002-45704 Supplier Medchemexpress LLC Supplier No. HY10177425MG
CSF1R-IN-1 is a potent small-molecule inhibitor of the colony stimulating factor 1 receptor (CSF1R) used for research into macrophage biology, tumor-associated macrophages, cancer, inflammation, and neurodegeneration. The compound has an IC50 of 0.5 nM against CSF1R, a molecular weight of 479.45 g/mol, and is supplied as a white to off-white solid with reported purity of 98.04%.
  • Potent CSF1R inhibition (IC50 0.5 nM).
  • High purity suitable for research (≈98.0%).
  • Defined molecular properties: C25H20F3N5O2, 479.45 g/mol.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage: powder -20°C (up to 3 years).
  • Available in small research quantities including 25 MG.