Learn More
Medchemexpress LLC HY-111787 10mg Medchemexpress, CSF1R-IN-2 CAS:2271119-26-5 Purity:>98%
SDP
Catalog No. 501968964
Encompass_Preferred
Medchemexpress, HY-111787 10mg CSF1R-IN-2 CAS:2271119-26-5 CSF1R-IN-2 (compound 5) is an oral-active inhibitor of SRC, MET and c-FMS, with IC50 values of 0.12 nM, 0.14 nM and 0.76 nM for SRC, MET and c-FMS respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8964 Supplier Medchemexpress LLC Supplier No. HY11178710MG
Medchemexpress, HY-111787 10mg CSF1R-IN-2 CAS:2271119-26-5 CSF1R-IN-2 (compound 5) is an oral-active inhibitor of SRC, MET and c-FMS, with IC50 values of 0.12 nM, 0.14 nM and 0.76 nM for SRC, MET and c-FMS respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.