Learn More
TARGETMOL CHEMICALS INC CSF1R-IN-3 5MG
SDP
Catalog No. 502579386
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CSF1R-IN-3 is an orally effective CSF-1R inhibitor with an IC50 value of 2.1 nM. CSF1R-IN-3 has anti-proliferative activity against colorectal cancer cells. CSF1R-IN-3 exhibits anti-tumour effects and immune enhancing effects by inhibiting the migration of macrophages and inducing the reprogramming of M2-type macrophages to M1-type macrophages thus exhibited activity against colorectal cancer cells. purity: 97%
Catalog No. 50-257-9386 Supplier TARGETMOL CHEMICALS INC Supplier No. T640665MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CSF1R-IN-3 is an orally effective CSF-1R inhibitor with an IC50 value of 2.1 nM. CSF1R-IN-3 has anti-proliferative activity against colorectal cancer cells. CSF1R-IN-3 exhibits anti-tumour effects and immune enhancing effects by inhibiting the migration of macrophages and inducing the reprogramming of M2-type macrophages to M1-type macrophages thus exhibited activity against colorectal cancer cells. purity: 97%