Learn More
Sino Biological CSF2RA / GM-CSFR / CD116 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10701MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD116 / GM-CSFR (rh CD116; Catalog#10701-H08H; P15509-1; Met 1-Gly 320). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography
Catalog No. 50-224-8544