Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1440MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CSF3/900. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: HL60 cells. Tonsil or lymph node. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human recombinant full-length CSF3 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 19kDa.Chromosome: 17q21.1. Gene Symbol: CSF3. Gene ID: 1440. Ensemble: ENSG00000108342. Uniprot: NP_001171618.
Catalog No. 50-188-4629