Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CSF3 (Human) ELISA Kit

Catalog No. 89100747 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-747 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-028-454 CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-747 Supplier Abnova™ Supplier No. KA3040
Add to Cart
Add to Cart

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CSF3

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description CSF3 (Human) ELISA Kit is intended for the quantitative measurement of human CSF3.
Detection Limit 4pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human G-CSF antibody, Lyophilized G-CSF standards (10ng x 2), Sample diluent buffer (30mL), Biotin conjugated anti-Human G-CSF antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (130uL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type CSF3 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.