Learn More
Velp™ CSF6 Dietary Fiber Extractor
Description
- Robust filtration unit for final filtration and washing phase defined in the enzymatic method for dietary fiber extraction
- Able to perform single or multiple samples at the same time for up to a maximum of six samples
- Highly efficient pump allows to speed-up filtration step and final washing
- Single or multiple sample processing
- Short filtration time under 20 minutes, even for six samples
- Electronic setting of counter pressure
Specifications
Specifications
| Product Type | Extractor |
| For Use With (Equipment) | CSF6 Filtration Unit |
| Frequency | 60 Hz |
| Voltage | 115 V |